Pipetti, yksikanavainen, kapillaari mikropipetti, Acura® capillar 846
Toimittaja: Socorex Isba
846.050 846.005 846.025 846.100 846.200
SOCO846.050EA 424 EUR
SOCO846.050 SOCO846.005 SOCO846.025 SOCO846.100 SOCO846.200
Pipetti, yksikanavainen, kapillaari mikropipetti, Acura® capillar 846
Pipetit ja pipetintäyttäjät Pipetit
High performance micropipettes with interchangeable glass capillary tube and ETFE tipped plunger.
  • Slim body shape, comfortable finger rest
  • Smooth plunger activation
  • Reliable, step-wise volume adjustment
  • Colour codes matching on pipettes, plungers and capillaries
  • Stainless steel, ETFE tipped plunger
The soft shaped handle and smooth plunger activation provide for enhanced hand comfort while handling viscous liquids, foaming solutions and volatile solvents.