Volumetric modules for Acura® electro
Toimittaja: Socorex Isba
800.0010 800.0050 800.0200 800.08.010 800.08.050 800.08.350 800.1000 800.10000 800.12.010 800.12.050 800.12.350 800.5000 800.1000XS 800.0200XS 800.0002 800.0002XS 800.0010Y 800.0020 800.0020XS 800.0050XS 800.0100XS 800.08.200 800.12.200 800.2000 800.0010YXS 800.0010XS 800.0100
635-0350EA 105 EUR
635-0350 635-0351 635-0352 635-0356 635-0357 635-0358 635-0353 635-0355 635-0359 635-0360 635-0361 635-0354 SOCO800.1000XS SOCO800.0200XS SOCO800.0002 SOCO800.0002XS SOCO800.0010Y SOCO800.0020 SOCO800.0020XS SOCO800.0050XS SOCO800.0100XS SOCO800.08.200 SOCO800.12.200 SOCO800.2000 SOCO800.0010YXS SOCO800.0010XS SOCO800.0100
Volumetric modules for Acura® electro
Pipetinkärjet
These interchangeable volumetric modules greatly extend working possibilities and make electronic pipetting affordable to all budgets. Disassembling and re-assembling without any tool contributes towards maximal flexibility.
Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.