Laboratory syringes, two ring, Dosys™ 160 series
Toimittaja: Socorex Isba
164.0501 164.0502 164.0505 164.0510 162.0501 163.05003 163.05005 163.0501 163.0502 162.0505 163.0510 163.0505 162.0502
SOCO164.0501EA 267 EUR
SOCO164.0501 SOCO164.0502 SOCO164.0505 SOCO164.0510 SOCO162.0501 SOCO163.05003 SOCO163.05005 SOCO163.0501 SOCO163.0502 SOCO162.0505 SOCO163.0510 SOCO163.0505 SOCO162.0502
Laboratory syringes, two ring, Dosys™ 160 series
Ruiskut Ruiskut, lääketieteellinen käyttö
Laboratory syringes are intended for safe and reliable liquid distribution, the selection of pistol-grip and two-ring instruments comes in a wide volume range.
  • Optimal hand balance and ergonomics
  • Fast and reliable volume setting
  • Plunger tightness without O-ring
  • High chemical resistance
Dosys™ 162 basic syringes are fill-by-fill (automatic) through needle or cannula, with luer lock nozzle.
Dosys™ 163 classic syringes are self-refilling automatic, with integrated valve system and luer lock nozzle.
Dosys™ 164 premium syringes are self-refillingMore Product Information