Laboratory syringes, Dosys™ 150 series
Toimittaja: Socorex Isba
155.0301 155.0305 155.0510 155.0505 155.0502 155.05100 155.05200 155.0520 155.05150 155.05250 155.0530 155.0550 155.0330 155.0320 155.03100 155.0310 155.0501
SOCO155.0301EA 31.1 EUR
SOCO155.0301 SOCO155.0305 SOCO155.0510 SOCO155.0505 SOCO155.0502 SOCO155.05100 SOCO155.05200 SOCO155.0520 SOCO155.05150 SOCO155.05250 SOCO155.0530 SOCO155.0550 SOCO155.0330 SOCO155.0320 SOCO155.03100 SOCO155.0310 SOCO155.0501
Laboratory syringes, Dosys™ 150 series
Ruiskut Ruiskut, lääketieteellinen käyttö
These reusable borosilicate glass syringes have great chemical and shock resistance and can be used for infusing, diluting, aspirating and sampling.
  • Excellent fitting and liquid tightness (not gas tight)
  • Permanent, high visibility graduations
  • Glass Luer or metal Luer lock nozzle