Replacement kits for Socorex® syringe
Toimittaja: Socorex Isba
1.187.72005 1.187.7201 1.187.7202 1.187.7205 1.187.7210 1.187.7220
SOCO1.187.72005EA 8.6 EUR
SOCO1.187.72005 SOCO1.187.7201 SOCO1.187.7202 SOCO1.187.7205 SOCO1.187.7210 SOCO1.187.7220
Replacement kits for Socorex® syringe
Venttiilit Injection Valves
Replacement kits for the valve assembly are supplied in a dedicated bag with each Socorex® self-refilling syringe.
Toimitustiedot: Each bag includes 2× springs, 2× balls, 2× washers for valve and 1× barrel washer.