Sterilin™ Petri Dishes
Toimittaja: Sterilin
502VF
SLIN502VFEA 576 EUR
SLIN502VF
Sterilin™ Petri Dishes
Soluviljelyalustat Petrimaljat
Ideal for use with different media or when savings in media or incubator space are required. Perform nonstandard applications with Thermo Scientific™ Sterilin™ Special Petri Dishes. Minimum order quantities apply. Not available in America.
  • Aseptically manufactured under clean room conditions (class 7 ISO14644) to exclude microbiological contamination
  • Single frosted marking area on the base of the dish - ideal for plate orientation with automatic pouring systems
  • CE marked in accordance with European Directive 98/79/EC
  • Manufactured to BS 611 part 2
  • In vitro use only
Order Now
TEKNISET TIEDOT
Environmentally Preferable