PCR racks for tubes, strips and plates, PCRack™
Toimittaja: Simport Scientific
T328-96AS T328-96B T328-96G T328-96O T328-96P T328-96R T328-96Y
SIMPT328-96ASEA 141 EUR
SIMPT328-96AS SIMPT328-96B SIMPT328-96G SIMPT328-96O SIMPT328-96P SIMPT328-96R SIMPT328-96Y
PCR racks for tubes, strips and plates, PCRack™
Koeputkitelineet Tube Racks
PCRack™ made of PP. Suitable for PCR individual tubes, 96 well plate and strips of 8 or 12 tubes.
  • Space saving
  • Stackable
It can be horizontally attached to each other in order to build-up any configuration. With the cover on, they are easily stackable one on top of another.