PCR-levyt, 96-kuoppaiset, korkeareunaiset, Amplate™
Toimittaja: Simport Scientific
T323-103N T323-100N T323-101N
SIMPT323-103NEA 423 EUR
SIMPT323-103N SIMPT323-100N SIMPT323-101N
PCR-levyt, 96-kuoppaiset, korkeareunaiset, Amplate™
Kuoppalevyt PCR-levyt
Amplate™ raised rim, thin walled PCR plates offer the rigidity needed for automation and are perfectly suited for high performance thermal cycling. Each well makes intimate contact with the heating block while quick and consistent heat transfer is ensured by a uniform wall thickness.
  • PP
  • Using polypropylene with low adhesion surface properties ensures samples are easily recovered
  • An alphanumeric grid helps sample identification
All sealing methods can be used for oil-free operation: Domed and flat cap strips; SecureSeal™ Thermal Sealing Film;More Product Information