Biopsy processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M526T
Toimittaja: Simport Scientific
M526-2T M526-3T M526-4T M526-5T M526-6T M526-7T M526-8T M526-9T M526-10T M526-11T M526-12T
SIMPM526-2TEA 266 EUR
SIMPM526-2T SIMPM526-3T SIMPM526-4T SIMPM526-5T SIMPM526-6T SIMPM526-7T SIMPM526-8T SIMPM526-9T SIMPM526-10T SIMPM526-11T SIMPM526-12T
Biopsy processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M526T
Histologiset kasetit Kudos- ja biopsiakasetit
These cassettes are made of acetal and are specially designed to hold biopsy specimens.
  • 1 mm square openings to maximise fluid exchange
  • Anterior writing area at a 45° angle
  • Taped cassettes to be used with Leica and Sakura ink jet printers
  • Easy to remove disposable cover
Unisette™ histology cassettes come with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
1 mm opening maximises fluid exchange and ensures proper drainage. The one-pieceMore Product Information