Kudos- ja biopsiakasetit QuickLoad™-telineissä, Swingsette™, M517T
Toimittaja: Simport Scientific
M517-4T M517-6T M517-2T M517-10T M517-11T M517-12T M517-3T M517-5T M517-7T M517-8T M517-9T
720-1982EA 544 EUR
720-1982 720-1983 SIMPM517-2T SIMPM517-10T SIMPM517-11T SIMPM517-12T SIMPM517-3T SIMPM517-5T SIMPM517-7T SIMPM517-8T SIMPM517-9T
Kudos- ja biopsiakasetit QuickLoad™-telineissä, Swingsette™, M517T
Histologiset kasetit Kasetit kudosten käsittelyyn
These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage. Suitable for use with Leica and Sakura labelers.
  • Made of acetal
  • Taped cassettes to be used with Leica and Sakura Ink Jet printers
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge