Viscosity flow cups
Toimittaja: Industrial Physics
VF2196 VF2197 VF2198 VF2195
SIMEVF2196EA 187 EUR
SIMEVF2196 SIMEVF2197 SIMEVF2198 SIMEVF2195
Viscosity flow cups
Viskosimetrit Viskosimetrit, viskositeettikuppimallit
These viscosity flow cups are titanium anodised aluminum viscosity cups.
Suitable to be used with stand, to measure the viscosity of lacquers, paint and other liquids.
Sertifikaatit: AFNOR