Conical bend testers
Toimittaja: Industrial Physics
SP1830 SP1831
SIMESP1830EA 1690 EUR
SIMESP1830 SIMESP1831
Conical bend testers
Pinnoitusten testauslaitteet
These Bend Test Mandrels are designed to bend coated panels over a conical shaped mandrel in order to assess the elasticity or resistance of a coating, paint or varnish to cracking, elongation and/or detachment from a metal test panel.
  • Sturdy apparatus in a combination of anodized aluminium and stainless steel
  • Large knob on bending arm for easy and smooth bending
  • Quick lock panel clamp (for Pro model only)
The 'Pro' model is secured by a quick lever handle, that locks and unlocks the panel, in a split second using just hand.
The 'Basic' model isMore Product Information