Pendulum hardness tester
Toimittaja: Industrial Physics
SP0510 SP0505 SP0500 SP0501
SIMESP0510EA 491 EUR
SIMESP0510 SIMESP0505 SIMESP0500 SIMESP0501
Pendulum hardness tester
Kovuusmittarit
Stainless steel.
  • Easy menu driven interface with jog dial
  • Design 'solid supports' to level the instrument
  • Integrated storage of glass plate and pendulum
  • Menu driven calibration menu eases both Persoz and König method
  • The end of a test is indicated by a visual and acoustic signal
The pendulum hardness test is based on the principle that the amplitude of oscillation of a pendulum touching a surface decreases more rapidly the softer the surface. It has a wide range of applications areas including laboratories, coating industry, paint industry, and theMore Product Information