Replica tapes, Press O-Film
Toimittaja: Industrial Physics
LD2071 LD2070 LD2067 LD2065 LD2066
SIMELD2071EA 85 EUR
SIMELD2071 SIMELD2070 SIMELD2067 SIMELD2065 SIMELD2066
Replica tapes, Press O-Film
Pinnoitusten testauslaitteet
This replica tape is specifically designed to measure blasted surfaces. By placing the Tqc replica tape on the surface and rubbing over it, the Rt (total roughness) or peak-to-valley height of the profile can be taken and measured.
Sertifikaatit: ASTM D4417-03