Combotester HI 9813 Multi-Parameter Tester (pH/Conductivity/TDS/°C), Handheld
Toimittaja: Industrial Physics
HI9813
SIMEHI9813EA 329 EUR
SIMEHI9813
Combotester HI 9813 Multi-Parameter Tester (pH/Conductivity/TDS/°C), Handheld
Sähkökemiamittarit pH-mittarit
The portable combo waterproof tester offer high accuracy pH, conductivity/TDS and temperature measurements in a single tester.
  • No more switching between meters for routine measurements
  • Waterproof, floating
  • Large, easy-to-read, dual-level LCD and automatic shut-off
  • pH and EC/TDS readings are automatically compensated for the effects of temperature (ATC)
  • EC/TDS conversion factor is user selectable as is the temperature compensation coefficient (ß)
This tester is supplied with a replaceable pH electrode cartridge with an extendable cloth junction as wellMore Product Information