Drying time recorder for film applicators
Toimittaja: Industrial Physics
AB3500
SIMEAB3500EA 801 EUR
SIMEAB3500
Drying time recorder for film applicators
Piirturit Piirturit
Drying time recorder tool can be used as an optional test tool for the automatic film applicators. This device allows the user to accurately define the drying time of various types of coatings within the time range of 1 minute to 48 hours. The drying time recorder option can be easily clamped on any standard automatic film applicator with firmware 2,01 and above.
  • With digital indication on drying stage
  • 'Intelligent recording' option significantly improves the resolution of longer drying process