Villanueva Osteochrome bone stain
Toimittaja: SIGMA ALDRICH MICROSCOPY
Vaara
SHH0027-450ML
SHH0027-450MLEA 270 EUR
SHH0027-450ML
Villanueva Osteochrome bone stain
Villanueva Osteochrome Bone stain
Useful for staining fresh, fixed, unembedded or plastic-embedded sections of bone. Villanueva osteochrome bone stain gives uniform and reproducible results. Useful in studying biopsy or postmortem tissue.