Acrylamide : Bisacrylamide (19:1) 40% in aqueous solution

Toimittaja: IBI SCIENTIFIC
Vaara

Synonyms: Acrylamide-Bis

IB70014
SHETIB70014EA 394 EUR
SHETIB70014
Acrylamide : Bisacrylamide (19:1) 40% in aqueous solution
Acrylamide : Bisacrylamide
Tiheys: 1.03 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00080848
UN: 3426
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR