Human/Mouse Recombinant TGF-B 3 (from E. coli)
Toimittaja: Shenandoah Biotechnology
800-11AF-100UG 800-11AF-10UG 800-11AF-1MG 800-11AF-500UG
SHBT800-11AF-100UGEA 778 EUR
SHBT800-11AF-100UG SHBT800-11AF-10UG SHBT800-11AF-1MG SHBT800-11AF-500UG
Human/Mouse Recombinant TGF-B 3 (from E. coli)
Proteiinit
Transforming growth factors (TGFs) are multifunctional peptides that regulate growth and differentiation in most cell types. The TGF-β family of proteins signal through serine/threonine kinase receptors. TGF-β isoforms (TGF-β1, -β2, and –β3) have overlapping, yet distinct biological actions in developing and adult tissues. TGF-β3 is an important factor in regulating cell adhesion and accelerating wound repair. TGF-β3 also functions during osteoblast proliferation, chemotaxis, and collagen synthesis.
  • High quality
  • Low endotoxin
  • Affordable
Varoitus: This product is for research use only and is not for use in humans.