Human/Mouse Recombinant TGF-B 3 (from E. coli)
Toimittaja: Shenandoah Biotechnology
800-11-100UG 800-11-10UG 800-11-1MG 800-11-500UG
SHBT800-11-100UGEA 705 EUR
SHBT800-11-100UG SHBT800-11-10UG SHBT800-11-1MG SHBT800-11-500UG
Human/Mouse Recombinant TGF-B 3 (from E. coli)
Proteiinit
Transforming growth factors (TGFs) are multifunctional peptides that regulate growth and differentiation in most cell types. The TGF-β family of proteins signal through serine/threonine kinase receptors. TGF-β isoforms (TGF-β1, -β2, and –β3) have overlapping, yet distinct biological actions in developing and adult tissues. TGF-β3 is an important factor in regulating cell adhesion and accelerating wound repair. TGF-β3 also functions during osteoblast proliferation, chemotaxis, and collagen synthesis.
  • High quality
  • Low endotoxin
  • Affordable
Varoitus: For research use only.