Mouse Recombinant IL-31 (from E. coli)
Toimittaja: Shenandoah Biotechnology
200-71AF-100UG 200-71AF-10UG 200-71AF-1MG 200-71AF-500UG
SHBT200-71AF-100UGEA 699 EUR
SHBT200-71AF-100UG SHBT200-71AF-10UG SHBT200-71AF-1MG SHBT200-71AF-500UG
Mouse Recombinant IL-31 (from E. coli)
Proteiinit
Interleukin 31 (IL-31) is an immunoregulatory cytokine that is expressed by activated type 2 T helper (Th2) cells. IL-31 signals through a heterodimer receptor consisting of the IL-31 Receptor A (IL-31RA) and the oncostatin M receptor (OSMR), which are expressed on monocytes, epithelial cells, and keratinocytes. IL-31 promotes allergic reactions and inflammatory skin diseases.
  • High quality
  • Low endotoxin
  • Affordable
Varoitus: For research use only.