Human Recombinant PLGF-3 (from E. coli)
Toimittaja: Shenandoah Biotechnology
100-198AF-100U 100-198AF-1MG 100-198AF-25UG 100-198AF-500U 100-198AF-5UG
SHBT100-198AF-100UEA 645 EUR
SHBT100-198AF-100U SHBT100-198AF-1MG SHBT100-198AF-25UG SHBT100-198AF-500U SHBT100-198AF-5UG
Human Recombinant PLGF-3 (from E. coli)
Proteiinit
Placenta growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family and promotes endothelial cell growth and angiogenesis. PlGF is secreted as a homodimer and may also form a heterodimer with VEGF. PlGF is detected in the placenta, heart, lungs, thyroid, and adipose tissues. Circulating PlGF levels are correlated with colorectal and renal cancers, as well as atherosclerosis and ischemic heart disease. There are four alternatively spliced PlGF isoforms (PlGF-1, PlGF-2, PlGF-3, and PlGF-4), each with unique secretion patterns and heparin-binding affinities. PlGF-3 lacks a heparin-binding domain and signals through the VEGFR-1 receptor.
  • High quality
  • Low endotoxin
  • Affordable
Varoitus: For research use only.