Filter cards, brown, for TPX chambers, SHANDON™
Toimittaja: Epredia
5991023
SHAN5991023EA 88.9 EUR
SHAN5991023
Filter cards, brown, for TPX chambers, SHANDON™
Suodattimet Sentrifugisuodattimet
Highly absorbent disposable filter cards for use with TPX chamber.
  • Use for sample volumes of 0,4 ml or less
  • For single chambers
Not for use with Cytofunnel™ disposable sample chambers.