Microtome blades, Shandon
Toimittaja: Epredia
3150734 3050822 3051835 3052835
SHAN3150734EA 413 EUR
SHAN3150734 SHAN3050822 SHAN3051835 720-2791
Microtome blades, Shandon
Mikrotomit ja mikrotomin terät Microtome Blades and Handles
Choose from HP35, MB22, or MX35 microtome blades.
  • Blades are available in high- and low-profile configurations
The MB22’s steeper angle allows users to accurately cut thinner sections faster.
HP35 disposable microtome blades for routine applications with a high proportion of harder tissue.
High-performance MX35 Premier disposable low-profile microtome blades for sectioning hard tissues and high-quality thin sections. Premier Plus blades provide greater durability, superior sharpness for lower blade friction and longer blade life.