HyClone Cell Boost™ Process Supplement Kit
Toimittaja: HyClone products (Cytiva)
SH30890
SH30890EA 857 EUR
SH30890
HyClone Cell Boost™ Process Supplement Kit
Soluviljelyyn suplementit ja lisäaineet
The HyClone Cell Boost™ process supplement kit contains 6×100 g samples of supplements designed to increase cell productivity in a variety of cell lines. Each supplement is designed to provide nutrients to fed-batch culture applications.
  • Supplements are chemically-defined and contain no animal components
  • Supplements have been tested in a variety of cell culture systems, including shaker flasks and bioreactors
  • Each supplement is also available in larger packaging configurations, please enquire for further information
Each Cell Boost formula is designed to provide a range of nutrients in novel combinations for multiple mammalian cell types, ideal for fed-batch applications or media customization. Each supplement has been tested in a variety of cell culture systems, including shaker flasks and bioreactors. 