Human mesenchymal stem cell screened fetal bovine serum (FBS), U.S. origin, Hyclone™
Toimittaja: HyClone products (Cytiva)
SH30071.03M
SH30071.03MEA 704 EUR
SH30071.03M
Human mesenchymal stem cell screened fetal bovine serum (FBS), U.S. origin, Hyclone™
Seerumit
Enhance human mesenchymal stem cell growth.
  • Designed for human mesenchymal stem cells
  • Sourced in the USA with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113,53
  • 40 nm filtration
Each lot is screened by growing human mesenchymal stem cells (hMSC) over a minimum of three subculture generations with 10 percent serum. Human MSC are then observed for evidence of nutritional deficiency, cytotoxicity or morphological aberrations. The serum is tested in parallel with aMore Product Information