L-MARK® LC Autosampler Syringes
Toimittaja: Trajan Scientific and Medical
LMK.3030031 LMK.2616704C LMK.2606416-6P LMK.2620719 LMK.2616502C LMK.2620720-6P LMK.2620614 LMK.2620935-BL LMK.2620718-2P LMK.2620518 LMK.2620517-2P LMK.2620720C LMK.3030022 LMK.2620617-2P LMK.2620615-6P LMK.3030021 LMK.2616422 LMK.2620718 LMK.2620517BL LMK.3030095 LMK.2606416-2P LMK.2620735-BL LMK.2620820-6P LMK.2620616-2P LMK.2620721-2P LMK.2620719-BL LMK.2620517 LMK.2620616 LMK.2620616-6P LMK.2620721-6P LMK.2620719-2P LMK.2620719-6P LMK.2620718C LMK.2620518C LMK.2620718-6P LMK.2620821-6P LMK.2620720-2P LMK.2620517C KLMK.26208212P LMK.2616822 LMK.2620721C LMK.2616922 LMK.2620721 KLMK.2620821 LMK.2620720 LMK.2620517-6P LMK.2606416 LMK.2620615-2P LMK.2620617-6P LMK.3030908 LMK.2620080-BL LMK.2620521C LMK.2620615 LMK.2620820-2P LMK.2620835-BL LMK.3030029 LMK.2620719C LMK.2620518-6P LMK.2620614C LMK.2620820 LMK.2620617 LMK.2620617C LMK.2606416C LMK.2620740 LMK.2620614-2P LMK.2620615C LMK2620617C-BL LMK2620721C-BL LMK2606416C-6P LMK2620517C-6P LMK2620615C-BL LMK2620719C-6P LMK2620721C-6P LMK2620517C-BL LMK2620719C-BL LMK2620719C-2P LMK2620719B-6P LMK2606416B-6P LMK2620821C-6P LMK2620616C-6P
SGEALMK.3030031EA 139 EUR
SGEALMK.3030031 SGEALMK.2616704C SGEALMK.2606416-6P SGEALMK.2620719 SGEALMK.2616502C SGEALMK.2620720-6P SGEALMK.2620614 SGEALMK.2620935-BL SGEALMK.2620718-2P SGEALMK.2620518 SGEALMK.2620517-2P SGEALMK.2620720C SGEALMK.3030022 SGEALMK.2620617-2P SGEALMK.2620615-6P SGEALMK.3030021 SGEALMK.2616422 SGEALMK.2620718 SGEALMK.2620517BL SGEALMK.3030095 SGEALMK.2606416-2P SGEALMK.2620735-BL SGEALMK.2620820-6P SGEALMK.2620616-2P SGEALMK.2620721-2P SGEALMK.2620719-BL SGEALMK.2620517 SGEALMK.2620616 SGEALMK.2620616-6P SGEALMK.2620721-6P SGEALMK.2620719-2P SGEALMK.2620719-6P SGEALMK.2620718C SGEALMK.2620518C SGEALMK.2620718-6P SGEALMK.2620821-6P SGEALMK.2620720-2P SGEALMK.2620517C SGEAKLMK.26208212P SGEALMK.2616822 SGEALMK.2620721C SGEALMK.2616922 SGEALMK.2620721 SGEAKLMK.2620821 SGEALMK.2620720 SGEALMK.2620517-6P SGEALMK.2606416 SGEALMK.2620615-2P SGEALMK.2620617-6P SGEALMK.3030908 SGEALMK.2620080-BL SGEALMK.2620521C SGEALMK.2620615 SGEALMK.2620820-2P SGEALMK.2620835-BL SGEALMK.3030029 SGEALMK.2620719C SGEALMK.2620518-6P SGEALMK.2620614C SGEALMK.2620820 SGEALMK.2620617 SGEALMK.2620617C SGEALMK.2606416C SGEALMK.2620740 SGEALMK.2620614-2P SGEALMK.2620615C SGEALMK2620617C-BL SGEALMK2620721C-BL SGEALMK2606416C-6P SGEALMK2620517C-6P SGEALMK2620615C-BL SGEALMK2620719C-6P SGEALMK2620721C-6P SGEALMK2620517C-BL SGEALMK2620719C-BL SGEALMK2620719C-2P SGEALMK2620719B-6P SGEALMK2606416B-6P SGEALMK2620821C-6P SGEALMK2620616C-6P
L-MARK® LC Autosampler Syringes
Ruiskut Ruiskut kromatografiaan
The comprehensive range of L-MARK® Gold and L-MARK syringes for LC systems are designed and tested to meet critical instrument parameters.
  • Plunger guide maintains optimal plunger alignment for extended plunger lifetime
  • Syringes available with polyethylene, PTFE and black PTFE tipped plungers
  • Titanium plungers for longer lifetime
  • Needle setting in the syringe barrel is protected by a PTFE barrier for reduced exposure of the adhesive to samples
  • The L-MARK Gold syringes include a chemically inert needle for extremely low carryover
Accuracy andMore Product Information