Pure-Pass™ Sealing Mats for Well Plates
Toimittaja: Trajan Scientific and Medical
462001 465001 465002 461001 461010
SGEA462001EA 46.3 EUR
SGEA462001 SGEA465001 SGEA465002 SGEA461001 SGEA461010
Pure-Pass™ Sealing Mats for Well Plates
Sulkijakalvot ja -matot Mikrolevyn tiivisteet
Pure-Pass sealing mats protect wells from leaks, contamination and evaporation during sampling and storage.
  • Available in non-slit and slit versions
  • Slit version enhances sample recovery by reducing vacuum formation
  • Plasma coating reduces the natural tackiness of silicone
These mats are available in EVA and specially processed low bleed, low extractable silicone to reduce sticking and a pre-slit version is available for easier piercing and sample withdrawal.
All mats are for 96 well plates with 7 mm diameter wells. Excellent dimensional conformanceMore Product Information