Pure-Pass™ Well Plates, 96- and 384-Wells
Toimittaja: Trajan Scientific and Medical
451001 451002 451006 451007 451008 451009 455001 455002 455006 455007 455008 455009
SGEA451001EA 85.8 EUR
SGEA451001 SGEA451002 SGEA451006 SGEA451007 SGEA451008 SGEA451009 SGEA455001 SGEA455002 SGEA455006 SGEA455007 SGEA455008 SGEA455009
Pure-Pass™ Well Plates, 96- and 384-Wells
Kuoppalevyt Syväkuoppalevyt
These as-molded, untreated plates are manufactured from a high purity polypropylene copolymer, making them some of the cleanest uncoated plastic plates.
Pure-Pass uncoated well plates are manufactured from a high clarity, high purity polypropylene copolymer for ultra-low extractables. These are molded in a cleanroon and meets all ANSI and SLAS standards.
Pure-Pass coated well plates with the addition of a glass barrier gives an added protection against extractables. Surface chemistry like glass could significantly reduce adsorption risk.