Vespel® Graphite GC Ferrules
Toimittaja: Trajan Scientific and Medical
213100 213103 213104 213105 213106 213108 213110 213124 213125 213163 213164 213165 213168 2131XX 213200 213204 213205 213208 213210 213222 213228 2132XX 213400 213405 213406 213408 213410 213420 2134XX 213800 215100 215103 215104 215105 215106 215108 215110 215124 215125 215164 215165 215168 2151XX 215200 215204 215205 215208 215210 2152XX 215400 2154XX 213404 213400B 215400B
SGEA213100EA 59.6 EUR
SGEA213100 SGEA213103 SGEA213104 SGEA213105 SGEA213106 SGEA213108 SGEA213110 SGEA213124 SGEA213125 SGEA213163 SGEA213164 SGEA213165 SGEA213168 SGEA2131XX SGEA213200 SGEA213204 SGEA213205 SGEA213208 SGEA213210 SGEA213222 SGEA213228 SGEA2132XX SGEA213400 SGEA213405 SGEA213406 SGEA213408 SGEA213410 SGEA213420 SGEA2134XX SGEA213800 SGEA215100 SGEA215103 SGEA215104 SGEA215105 SGEA215106 SGEA215108 SGEA215110 SGEA215124 SGEA215125 SGEA215164 SGEA215165 SGEA215168 SGEA2151XX SGEA215200 SGEA215204 SGEA215205 SGEA215208 SGEA215210 SGEA2152XX SGEA215400 SGEA2154XX SGEA213404 SGEA213400B SGEA215400B
Vespel® Graphite GC Ferrules
Kiinnitykset ja liittimet
Vespel/graphite GC ferrules are made from quality polyimide/graphite blend. They seal with minimal torque and can be reused and repositioned along capillary columns if not over compressed in the fitting. Due to slight shrinking at high temperatures, they must be retightened after initial temperature cycles to avoid leaks.
  • Trajan offers both Vespel/graphite blends, a 60/40 mix and an 85/15 mix
  • The 85/15 maintains a high level of hardness and long lifetime
  • The 60/40 blend is softer, with improved wear resistance and lower friction
  • These ferrules will require