CRS ZPure™ Hydrocarbon Gas Filters
Toimittaja: Trajan Scientific and Medical
202232-B 202232D-B 202232D-SS 202232L-B 202232L-SS 202232-SS 202232XL-B 202232XL-SS 202233-B 202233D-B 202233D-SS 202233L-B 202233L-SS 202233-SS 202233XL-B 202233XL-SS 202232-4-PC 202232-6-PC 202232-B-QC 202232D-4-PC 202232D-6-PC 202232D-B-QC 202232D-SS-QC 202232L-4-PC 202232L-6-PC 202232L-B-QC 202232L-SS-QC 202232-SS-QC 202232XL-4-PC 202232XL-6-PC 202232XL-B-QC 202232XL-SS-QC 202233-B-QC 202233D-B-QC 202233D-SS-QC 202233L-B-QC 202233L-SS-QC 202233-SS-QC 202233XL-B-QC 202233XL-SS-QC 202RHC-B-QC 202RHCD-B-QC 202RHCD-SS-QC 202RHCL-B-QC 202RHCL-SS-QC 202RHC-SS-QC 202RHCXL-B-QC 202RHCXL-SS-QC
SGEA202232-BEA 196 EUR
SGEA202232-B SGEA202232D-B SGEA202232D-SS SGEA202232L-B SGEA202232L-SS SGEA202232-SS SGEA202232XL-B SGEA202232XL-SS SGEA202233-B SGEA202233D-B SGEA202233D-SS SGEA202233L-B SGEA202233L-SS SGEA202233-SS SGEA202233XL-B SGEA202233XL-SS SGEA202232-4-PC SGEA202232-6-PC SGEA202232-B-QC SGEA202232D-4-PC SGEA202232D-6-PC SGEA202232D-B-QC SGEA202232D-SS-QC SGEA202232L-4-PC SGEA202232L-6-PC SGEA202232L-B-QC SGEA202232L-SS-QC SGEA202232-SS-QC SGEA202232XL-4-PC SGEA202232XL-6-PC SGEA202232XL-B-QC SGEA202232XL-SS-QC SGEA202233-B-QC SGEA202233D-B-QC SGEA202233D-SS-QC SGEA202233L-B-QC SGEA202233L-SS-QC SGEA202233-SS-QC SGEA202233XL-B-QC SGEA202233XL-SS-QC SGEA202RHC-B-QC SGEA202RHCD-B-QC SGEA202RHCD-SS-QC SGEA202RHCL-B-QC SGEA202RHCL-SS-QC SGEA202RHC-SS-QC SGEA202RHCXL-B-QC SGEA202RHCXL-SS-QC
CRS ZPure™ Hydrocarbon Gas Filters
Suodattimet Kaasusuodattimet
The ZPure™ line of filters remove a wide range of contaminants to trace levels.
  • High-efficiency in-line traps with outstanding capacity
  • High quality activated absorbents for long purifier life and efficient contaminant removal
  • Various size and fitting configurations to fit existing installations
  • Filter lifetime is dependent on quality of incoming gas, and the flow rate
  • Individually leak-tested
Applications: It can be used to remove hydrocarbons heavier than butane from inert gases, He, Ar, N₂, H₂, and clean dry air (CDA) to low ppb levels. It isMore Product Information