CRS ZPure™ Ultra Gas Filters
Toimittaja: Trajan Scientific and Medical
202371-V 202373-V 202375-V
SGEA202371-VEA 600 EUR
SGEA202371-V SGEA202373-V SGEA202375-V
CRS ZPure™ Ultra Gas Filters
Suodattimet Kaasusuodattimet
The ZPure™ ultra filters are designed for use when extremely pure gas is critical to the application such as in the microelectronics and the semiconductors industry.
  • Made up of high quality 316 stainless steel
  • Electro polished surface finish
  • outlet purity to low ppb levels
  • for use at ambient temperatures
  • Available in 3A or 5A moisture removing