CRS ZPure™ PolyGas Gas Purifiers
Toimittaja: Trajan Scientific and Medical
202285-B 202285D-B 202285D-SS 202285L-B 202285L-SS 202285-SS 202285XL-B 202285XL-SS 202286-B 202286D-B 202286D-SS 202286L-B 202286L-SS 202286-SS 202286XL-B 202286XL-SS 202287-B 202287D-B 202287D-SS 202287L-B 202287L-SS 202287-SS 202287XL-B 202287XL-SS 202288-B 202288D-B 202288D-SS 202288L-B 202288L-SS 202288-SS 202288XL-B 202288XL-SS 202285-B-QC 202285D-B-QC 202285D-SS-QC 202285L-B-QC 202285L-SS-QC 202285-SS-QC 202285XL-B-QC 202285XL-SS-QC 202286-B-QC 202286D-B-QC 202286D-SS-QC 202286L-B-QC 202286L-SS-QC 202286-SS-QC 202286XL-B-QC 202286XL-SS-QC 202287-B-QC 202287D-B-QC 202287D-SS-QC 202287L-B-QC 202287L-SS-QC 202287-SS-QC 202287XL-B-QC 202287XL-SS-QC 202288-B-QC 202288D-B-QC 202288D-SS-QC 202288L-B-QC 202288L-SS-QC 202288-SS-QC 202288XL-B-QC 202288XL-SS-QC 202RP1-B-QC 202RP1D-B-QC 202RP1D-SS-QC 202RP1L-B-QC 202RP1L-SS-QC 202RP1-SS-QC 202RP1XL-B-QC 202RP1XL-SS-QC 202RP2-B-QC 202RP2D-B-QC 202RP2D-SS-QC 202RP2L-B-QC 202RP2L-SS-QC 202RP2-SS-QC 202RP2XL-B-QC 202RP2XL-SS-QC
SGEA202285-BEA 268 EUR
SGEA202285-B SGEA202285D-B SGEA202285D-SS SGEA202285L-B SGEA202285L-SS SGEA202285-SS SGEA202285XL-B SGEA202285XL-SS SGEA202286-B SGEA202286D-B SGEA202286D-SS SGEA202286L-B SGEA202286L-SS SGEA202286-SS SGEA202286XL-B SGEA202286XL-SS SGEA202287-B SGEA202287D-B SGEA202287D-SS SGEA202287L-B SGEA202287L-SS SGEA202287-SS SGEA202287XL-B SGEA202287XL-SS SGEA202288-B SGEA202288D-B SGEA202288D-SS SGEA202288L-B SGEA202288L-SS SGEA202288-SS SGEA202288XL-B SGEA202288XL-SS SGEA202285-B-QC SGEA202285D-B-QC SGEA202285D-SS-QC SGEA202285L-B-QC SGEA202285L-SS-QC SGEA202285-SS-QC SGEA202285XL-B-QC SGEA202285XL-SS-QC SGEA202286-B-QC SGEA202286D-B-QC SGEA202286D-SS-QC SGEA202286L-B-QC SGEA202286L-SS-QC SGEA202286-SS-QC SGEA202286XL-B-QC SGEA202286XL-SS-QC SGEA202287-B-QC SGEA202287D-B-QC SGEA202287D-SS-QC SGEA202287L-B-QC SGEA202287L-SS-QC SGEA202287-SS-QC SGEA202287XL-B-QC SGEA202287XL-SS-QC SGEA202288-B-QC SGEA202288D-B-QC SGEA202288D-SS-QC SGEA202288L-B-QC SGEA202288L-SS-QC SGEA202288-SS-QC SGEA202288XL-B-QC SGEA202288XL-SS-QC SGEA202RP1-B-QC SGEA202RP1D-B-QC SGEA202RP1D-SS-QC SGEA202RP1L-B-QC SGEA202RP1L-SS-QC SGEA202RP1-SS-QC SGEA202RP1XL-B-QC SGEA202RP1XL-SS-QC SGEA202RP2-B-QC SGEA202RP2D-B-QC SGEA202RP2D-SS-QC SGEA202RP2L-B-QC SGEA202RP2L-SS-QC SGEA202RP2-SS-QC SGEA202RP2XL-B-QC SGEA202RP2XL-SS-QC
CRS ZPure™ PolyGas Gas Purifiers
Suodattimet Kaasusuodattimet
The ZPure PolyGas purifiers removes a wide range of contaminants including O₂, H₂O, CO₂, CO, halogenated compounds, siloxanes, hydrocarbons heavier than butane, sulfur-containing molecules,
ammonia and other gases.
  • The ZPure PolyGas purifier can be used to purify inert gases, He, Ar, N₂ and H₂
The ZPure PolyGas I purifier is ideal for use with GC and GC/MS carrier gas lines. It is also recommended for any application requiring ultra-pure gas free from oxygen, water and hydrocarbons (heavier than butane).
The ZPure PolyGas II purifier is used toMore Product Information