CRS ZPure™ H₂O HSMI Indicating Filters
Toimittaja: Trajan Scientific and Medical
202283-B 202283-SS 202284-B 202284-SS 202283-B-QC 202283RP-B-QC 202283RP-SS-QC 202283-SS-QC 202284-B-QC 202284-SS-QC
SGEA202283-BEA 236 EUR
SGEA202283-B SGEA202283-SS SGEA202284-B SGEA202284-SS SGEA202283-B-QC SGEA202283RP-B-QC SGEA202283RP-SS-QC SGEA202283-SS-QC SGEA202284-B-QC SGEA202284-SS-QC
CRS ZPure™ H₂O HSMI Indicating Filters
Suodattimet Kaasusuodattimet
The ZPure™ H₂O 'High Sensitivity Moisture Indicating' Filter is used for applications requiring high efficiency moisture removal. The high sensitivity indicator signals breakthrough at 1 ppm water and changes from blue to brown.
  • High-efficiency in-line traps with outstanding capacity
  • High-quality activated adsorbents for long purifier life
  • Filter lifetime is dependent of quality of incoming gas and the flow rate
  • Highly sensitive indicators
  • Double safety seal and individually leak-tested
It can be used to purify inert gases, He, Ar, N₂ and H₂ for anyMore Product Information