CRS ZPure™ Glass H₂O/HC Filters
Toimittaja: Trajan Scientific and Medical
202256-B 202256-B-QC 202256RP-B-QC 202256RP-SS-QC 202256-SS 202256-SS-QC 202257-B 202257-B-QC 202257-SS 202257-SS-QC
SGEA202256-BEA 216 EUR
SGEA202256-B SGEA202256-B-QC SGEA202256RP-B-QC SGEA202256RP-SS-QC SGEA202256-SS SGEA202256-SS-QC SGEA202257-B SGEA202257-B-QC SGEA202257-SS SGEA202257-SS-QC
CRS ZPure™ Glass H₂O/HC Filters
Suodattimet Kaasusuodattimet
The ZPure™ H₂O/HC filter removes moisture and hydrocarbons from dry air or inert gases. The co-free moisture indicator signals breakthrough when it changes from green to blue.
Dual seal design: In this design the heavy-walled inner glass tube is protected by a sealed outer plastic tube. This design provides extra protection if the glass tube were to fail, the gas system is still fully protected against leaks. An internal frit at each end prevents particulate contamination.
The ZPure™ Glass H₂O/HC Filter is available with standard compression fittings or quick connect fittings in ¹/₄"More Product Information