CRS ZPure™ M Gas Filters
Toimittaja: Trajan Scientific and Medical
202230M-V 202231M-SS 202232M-V 202233M-SS 202287M-V 202288M-SS 202295M-V 202296M-SS 202231M-SS-QC 202233M-SS-QC 202288M-SS-QC 202296M-SS-QC
SGEA202230M-VEA 4280 EUR
SGEA202230M-V SGEA202231M-SS SGEA202232M-V SGEA202233M-SS SGEA202287M-V SGEA202288M-SS SGEA202295M-V SGEA202296M-SS SGEA202231M-SS-QC SGEA202233M-SS-QC SGEA202288M-SS-QC SGEA202296M-SS-QC
CRS ZPure™ M Gas Filters
Suodattimet Kaasusuodattimet
The ZPure™ M filters remove a wide range of contaminants to trace levels. The ZPure™ M filters are 3800 cc canisters that have the benefits of higher flow rates, larger capacities and are refillable in most versions.
  • High efficiency in-line traps with outstanding capacity
  • High quality activated adsorbents for long purifier life and efficient contaminant removal
  • Filter lifetime is dependent on quality of incoming gas and the flow rate
  • Made from 316 stainless steel
  • Individually leak tested
Application: It can be used to purify inert gases such as He, Ar,More Product Information