CRS ZPure™ CO₂ Gas Filters
Toimittaja: Trajan Scientific and Medical
202212-4-PC 202212-6-PC 202212D-4-PC 202212D-6-PC 202RCO-B-QC 202RCOD-B-QC 202RCOD-SS-QC 202RCOL-B-QC 202RCOL-SS-QC 202RCO-SS-QC 202RCOXL-B-QC 202RCOXL-SS-QC
SGEA202212-4-PCEA 246 EUR
SGEA202212-4-PC SGEA202212-6-PC SGEA202212D-4-PC SGEA202212D-6-PC SGEA202RCO-B-QC SGEA202RCOD-B-QC SGEA202RCOD-SS-QC SGEA202RCOL-B-QC SGEA202RCOL-SS-QC SGEA202RCO-SS-QC SGEA202RCOXL-B-QC SGEA202RCOXL-SS-QC
CRS ZPure™ CO₂ Gas Filters
Suodattimet Kaasusuodattimet
The ZPure CO₂ filter removes carbon dioxide from inert gases, He, Ar, N₂, H₂, and clean dry air (CDA) to low ppb levels.
The filter functions by consuming carbon dioxide in a reaction with highly-dispersed NaOH on a silicate support. The reaction produces water which remains adsorbed in the purifier. In highly critical applications consideration should be given to using an additional moisture trap after the CO₂ filter as a safety measure.
It is available in 4 sizes of stainless steel cylinders ranging from 130 to 750 ml. The ZPure CO₂ filter is available with standard compressionMore Product Information