Gas purifiers/filters, Big Trap
Toimittaja: Trajan Scientific and Medical
1035338 1035334 1035328 1035324 1035318 1035314 1035348 1035344
SGEA1035338EA 398 EUR
SGEA1035338 SGEA1035334 SGEA1035328 SGEA1035324 SGEA1035318 SGEA1035314 SGEA1035348 SGEA1035344
Gas purifiers/filters, Big Trap
Suodattimet Kaasusuodattimet
Big Trap gas purifiers are designed for bulk purification applications, or where several instruments are plumbed from a single source.
  • One-piece, heavy walled aluminium tube
  • Capacity: 750 cm³
  • Pressure rating up to 17,2 bar
Gas purifiers are an essential part of your GC analysis as contaminants in gases can significantly impact the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation. Even if carrier gas is of the highest quality,More Product Information