SGE OptChem™ Single Taper Inlet Liners
Toimittaja: Trajan Scientific and Medical
0926201701 09262017 0926201501 09262015
SGEA0926201701EA 58.2 EUR
SGEA0926201701 SGEA09262017 SGEA0926201501 SGEA09262015
SGE OptChem™ Single Taper Inlet Liners
Kromatografiaan tarvikkeet Kromatografiaan läpivientiholkki
SGE OptChem™ GC inlet liner provide optimal performance with advanced coating technologies in an easy to use format for GC and GCMS.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - ready to install touchless packaging with pre-fitted CRS ONE O-rings
  • OptChem™ deactivation is a unique thick film which limits breakdown, ensuring minimal loss of sample and increased system sensitivity
  • Touchless packaging format reduces the risk of contamination during installation
  • Recyclable packaging - less waste to landfill
  • Color coded