SGE OptChem™ FocusLiners Inlet Liners
Toimittaja: Trajan Scientific and Medical
0926200501 09262005 0926200201
SGEA0926200501EA 70.2 EUR
SGEA0926200501 SGEA09262005 SGEA0926200201
SGE OptChem™ FocusLiners Inlet Liners
Kromatografiaan tarvikkeet Kromatografiaan läpivientiholkki
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner. Choosing the right inlet liner and injection parameters can increase peak areas and reduce detection limits by up to 300%.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to use
  • Two focus points in the liner ensures quartz wool remains in the optimum location
  • The wiping of the needle by the quartz wool prevents droplet formation ensuring excellent reproducibility
  • The presence of the quartz