Jumbo syringes
Toimittaja: Trajan Scientific and Medical
009910 009920 009930
SGEA009910EA 1010 EUR
SGEA009910 SGEA009920 SGEA009930
Jumbo syringes
Ruiskut Ruiskut kromatografiaan
These syringes are ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
  • Designed for holding and dispensing large volumes of gas
  • Heavy duty acrylic barrels
  • Easy access to sample for the addition of standards or removal of subsample via secondary port
  • Plunger stem can be unscrewed for ease of transportation and storage