Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Toimittaja: VWR Seradigm
SERD76419-586EA 71.2 EUR
SERD76419-586 SERD76419-590 1300-500CR 1300-500CG 1300-500CGH 1300-500CH
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Seerumit
Avantor Seradigm Select Grade FBS provides superior performance and consistency in cost-sensitive applications.
  • Industry-leading collection techniques at the source
  • Risk mitigation through a robust quality system
  • Proven track record for performance and consistency
  • Sourced from countries with health status classified as “Negligible BSE Risk” by the World Health Organization
The care and attention we devote to the collection and manufacture of our products generates quality that matters. Avantor Seradigm Select Grade FBS preserves more of theMore Product Information