Käsineet, Semperguard® Xpert Powder-free
Toimittaja: SEMPERIT TECHN PROD
816780633 816780635 816780637 816780639
SEMP816780633EA 39.9 EUR
SEMP816780633 SEMP816780635 SEMP816780637 SEMP816780639
Käsineet, Semperguard® Xpert Powder-free
Käsineet
These disposable nitrile gloves for complex risks provide excellent resistance to skin irritants and chemicals. Suitable for use in medical environments, als well as in laboratories and for food processing.
  • Non sterile, powder-free, textured finish
  • More resilience through special formulation and greater wall thickness
  • Free from phthalates / softeners and allergenic latex proteins
  • Easy to don and particularly skin-friendly through special finishing process
  • Signal colour blue – suitable for areas subject to HACCP standards
Length: 240 mm
Thickness: 0,More Product Information