Käsineet, tutkimus, Semperguard®
Toimittaja: SEMPERIT TECHN PROD
813780043 813780047 813780045 813780041 813780049
SEMP813780043EA 22.7 EUR
SEMP813780043 SEMP813780047 SEMP813780045 SEMP813780041 SEMP813780049
Käsineet, tutkimus, Semperguard®
Käsineet
This disposable glove made of natural latex can be used in medical areas as well as in industry and in labs.
  • Non sterile
  • Particularly skin-friendly and powder-free
  • With synthetic coating on the inside and textured finish on the outside: Good grip and easy to don
  • Free from phthalates / softeners
  • Ambidextrous, with rolled edge
EN 374, EN 455, medical device class I, Cat. III; AQL 1,5 Food contact approved
Sertifikaatit: EN 374, EN 455, medical device class I, Cat. III; AQL 1,5
Food contact approved