Airless dispenser, ecosolution®
Toimittaja: SEMADENI
7501 7502
SEMA7501EA 434 EUR
SEMA7501 SEMA7502
Airless dispenser, ecosolution®
Pullot Aerosolisuihkepullot
Bottle made of polypropylene.
  • Contracts during dispensing and automatically ensures almost complete emptying of the content
  • Contents are air-protected
  • Dispenser pump is simply snapped on after filling
  • Ideal for viscous products such as lotions or creams
The dispenser pumps have a convenient shutter mechanism (¹/₄ turn to the left). Pumps are available in a short and long spout version (beak). The short version is equipped with an additional, transparent snap cover.
Volume per dosage (stroke): 1,5 ml