Jars with screw cap
Toimittaja: SEMADENI
5490 5491 5492 5496
SEMA5490EA 135 EUR
SEMA5490 SEMA5491 SEMA5492 SEMA5496
Jars with screw cap
Näyteastiat, muovia Yleiskäyttöinenastiat
Jars made of PP, white,with screw caps made of red PP.
  • Very good chemical resistance
  • Very good temperature resistance
Suitable for powders, granules, pastes or cosmetic products. Physiologically harmless and suitable for food storage.