Disposal canisters, with screw cap
Toimittaja: SEMADENI
2012 2823 2824 2013
216-7313EA 107 EUR
216-7313 SEMA2823 SEMA2824 SEMA2013
Disposal canisters, with screw cap
Pullot Jerry-kannut
HDPE, coloured.
  • Good chemical resistance
  • Good temperature resistance
  • With a hollow handle, filling opening and vent screw
  • Stackable
  • Neck Ø: 42 mm
For collecting chemicals.