Packaging and Storage Boxes
Toimittaja: SEMADENI
1734 1730 1733 3384 2966 3388 1982 1729 4677 1987 1717 1722 1943 4567 1738 1984 3386 3387 1986 2964 4560
215-0270EA 11.1 EUR
215-0270 216-3131 216-3133 216-3145 216-3147 216-3148 216-3157 SEMA1729 SEMA4677 SEMA1987 SEMA1717 SEMA1722 SEMA1943 SEMA4567 SEMA1738 SEMA1984 SEMA3386 SEMA3387 SEMA1986 SEMA2964 SEMA4560
Packaging and Storage Boxes
Laatikot Säilytyslaatikot
These boxes made of polystyrene (PS) are crystal clear with a good closing tight fitting lid.
  • Clear for easy product identification
  • The lids are either mounted or not mounted
  • Stackable