Recombinant human deiodinase 2

Toimittaja: SELENOZYME

HDIO2-100 HDIO2-200 HDIO2-500 HDIO2-1000
SELZHDIO2-100EA 700 EUR
SELZHDIO2-100 SELZHDIO2-200 SELZHDIO2-500 SELZHDIO2-1000
Recombinant human deiodinase 2
Proteiinit

The human DIO2 selenocprotein is encoded by the DIO2 gene and catalyzes the conversion of prohormone thyroxine (3,5,3',5'-tetraiodothyronine, T4) to the bioactive thyroid hormone (3,5,3'-triiodothyronine, T3) by outer ring 5'-deiodination.
We provide the human DIO2 enzyme with approx. 20% Sec contents, mixed with inactive forms containing Sec-to-Lys and Sec-to-Gln substitutions, at more than 95% purity in aliquots at amounts from 50 to 500 µg.


  • Genuine Sec-containing selenoproteins
  • More than 95% purity
  • No animals used as protein sources (grown in bacteria)


Toimitustiedot: Protein stored in TE buffer with 50% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.


Pakkaus: Aliquots

Order Now

TEKNISET TIEDOT

Environmentally Preferable
Lajeja
Lähde
Puhtaus ≥ 95%
Säilytysolosuhteet Protein stored in Tris-EDTA buffer with 50% glycerol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR