Safety Waste Caps, Electrically Conductive
Toimittaja: S.C.A.T.
460050 460060 460070 460090
SCTV460050EA 184 EUR
SCTV460050 SCTV460060 SCTV460070 SCTV460090
Safety Waste Caps, Electrically Conductive
Tulpat ja korkit Turvakorkit
SCAT Safety Waste Caps, made of electrically conductive PTFE, are hermetically sealing caps for the safe disposal of your liquid HPLC waste (e.g. solvents, acids, alkalis).
  • For the prevention of explosions due to static discharge
  • Made of electrically conductive PTFE material
  • Available in different thread sizes
The SCAT exhaust filter with three layers of activated carbon binds vapors. It thus additionally ensures safe pressure equalization and occupational safety.